EMAIL THIS PAGE TO A FRIEND
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V
Anticancer activity of galactoxyloglucan polysaccharide-conjugated doxorubicin nanoparticles: Mechanistic insights and interactome analysis.
PMID 25864443
Abstract
Toxicity associated with chemotherapeutic drugs such as doxorubicin (Dox), is one of the major obstacles that is currently affecting patients. PST-Dox (Galactoxyloglucan, PST001-conjugated Dox) nanoparticles were synthesized by encapsulating Dox with polysaccharide PST001, isolated from Tamarindus indica (Ti) by ionic gelation with tripolyphosphate (TPP). Herein, we demonstrate a detailed mechanistic and interactome network analysis that is specific to PST-Dox action in cancer cells and normal lymphocytes. Our results show that PST-Dox is superior to its parental counterparts, exhibiting a greater cytotoxicity by the induction of apoptosis against a wide variety of cancers by enhanced cellular uptake of Dox from the nanoparticle conjugates. Also, PST-Dox nanoparticles were non-toxic to normal lymphocytes with limited immunostimulatory effects up to certain doses. Elucidation of molecular mechanism by whole genome microarray in cancer cells and lymphocytes revealed that a large number of genes were dysregulated specifically in cancer cells. Specifically, a unique target gene EGR1, contextually determined translational activation of P53 in the cancerous and non-cancerous cells. Most of the key downregulated genes were tyrosine kinases, indicating the potential inhibitory action of PST-Dox on tyrosine kinase oncogenic pathways. Western blotting of proteins corresponding to the genes that were altered at the genomic level was very well correlated in the majority of them, except in a few that demonstrated post-transcriptional modifications. The important findings and highly disciplined approaches highlighted in the present study will speed up the therapeutic potential of this augmented nanoparticle formulation for more robust clinical studies and testing in several cancers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
28-4630
Sodium tripolyphosphate, SAJ first grade, 56.0-61.0% P2O5 basis
Na5O10P3