EMAIL THIS PAGE TO A FRIEND
Hippocampus
Cholinergic, but not NMDA, receptors in the lateral entorhinal cortex mediate acquisition in trace eyeblink conditioning.
PMID 25865030
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5282
DL-2-Amino-5-phosphonopentanoic acid, solid
C5H12NO5P
G019 Muscimol hydrobromide, ≥98% (HPLC), solid
C4H6N2O2 · HBr