EMAIL THIS PAGE TO A FRIEND
International journal of molecular sciences
Iron oxide nanoparticles for magnetically-guided and magnetically-responsive drug delivery.
PMID 25867479
Abstract
In this review, we discuss the recent advances in and problems with the use of magnetically-guided and magnetically-responsive nanoparticles in drug delivery and magnetofection. In magnetically-guided nanoparticles, a constant external magnetic field is used to transport magnetic nanoparticles loaded with drugs to a specific site within the body or to increase the transfection capacity. Magnetofection is the delivery of nucleic acids under the influence of a magnetic field acting on nucleic acid vectors that are associated with magnetic nanoparticles. In magnetically-responsive nanoparticles, magnetic nanoparticles are encapsulated or embedded in a larger colloidal structure that carries a drug. In this last case, an alternating magnetic field can modify the structure of the colloid, thereby providing spatial and temporal control over drug release.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
900148 Iron oxide (II,III), nanoparticles, 30 nm avg. part. size (TEM), streptavidin functionalized, 1 mg/mL in H2O
900062 Iron oxide (II,III), nanoparticles, 30 nm avg. part. size (TEM), 5 mg/mL in H2O
900043 Iron oxide (II,III), nanoparticles, 15 nm avg. part. size (TEM), 5 mg/mL in H2O
900091 Iron oxide (II,III), nanoparticles, 10 nm avg. part. size (TEM), streptavidin functionalized, 1 mg/mL in H2O
900084 Iron oxide (II,III), nanoparticles, 10 nm avg. part. size (TEM), 5 mg/mL in chloroform
900026 Iron oxide (II,III), nanoparticles, 15 nm avg. part. size (TEM), PEG functionalized, 1 mg/mL in H2O
900082 Iron oxide (II,III), nanoparticles, 5 nm avg. part. size (TEM), 5 mg/mL in chloroform
900042 Iron oxide (II,III), nanoparticles, 25 nm avg. part. size (TEM), 5 mg/mL in H2O
900093 Iron oxide (II,III), nanoparticles, 20 nm avg. part. size (TEM), streptavidin functionalized, 1 mg/mL in H2O
900063 Iron oxide (II,III), nanoparticles, 15 nm avg. part. size (TEM), 5 mg/mL in toluene
900028 Iron oxide (II,III), nanoparticles, 25 nm avg. part. size (TEM), amine functionalized, 1 mg/mL in H2O
900201 Iron oxide (II,III), nanoparticles, 25 nm avg. part. size (TEM), carboxylic acid functionalized, 5 mg/mL in H2O
900199 Iron oxide (II,III), nanoparticles, 15 nm avg. part. size (TEM), amine functionalized, 1 mg/mL in H2O
900200 Iron oxide (II,III), nanoparticles, 15 nm avg. part. size (TEM), carboxylic acid functionalized, 5 mg/mL in H2O
900146 Iron oxide (II,III), nanoparticles, 10 nm avg. part. size (TEM), rhodamine B functionalized, 1 mg/mL in H2O
900147 Iron oxide (II,III), nanoparticles, 5 nm avg. part. size (TEM), dextran functionalized, 10 mg/mL in H2O
900088 Iron oxide (II,III), nanoparticles, 20 nm avg. part. size (TEM), 5 mg/mL in chloroform
900089 Iron oxide (II,III), nanoparticles, 25 nm avg. part. size (TEM), 5 mg/mL in chloroform
900090 Iron oxide (II,III), nanoparticles, 30 nm avg. part. size (TEM), 5 mg/mL in chloroform
900092 Iron oxide (II,III), nanoparticles, 15 nm avg. part. size (TEM), streptavidin functionalized, 1 mg/mL in H2O
900094 Iron oxide (II,III), nanoparticles, 25 nm avg. part. size (TEM), streptavidin functionalized, 1 mg/mL in H2O
900083 Iron oxide (II,III), nanoparticles, 5 mg/mL in chloroform
900037 Iron oxide (II,III), nanoparticles, 15 nm avg. part. size (TMS), 1 mg/mL (in 10 mM PBS buffer), biotin functionalized
900081 Iron oxide (II,III), nanoparticles, 30 nm avg. part. size (TEM), 5 mg/mL in toluene
900064 Iron oxide (II,III), nanoparticles, 25 nm avg. part. size (TEM), 5 mg/mL in toluene
900027 Iron oxide (II,III), nanoparticles, 25 nm avg. part. size (TEM), PEG functionalized, 1 mg/mL in H2O
900034 Iron oxide (II,III), nanopowder, 25 nm, NHS ester functionalized
900041 Iron oxide (II,III), nanopowder, 15 nm avg. part. size (TEM), NHS ester functionalized