EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
Functional interplay between the RK motif and linker segment dictates Oct4-DNA recognition.
PMID 25870414
Abstract
The POU family transcription factor Oct4 plays pivotal roles in regulating pluripotency and somatic cell reprogramming. Previous studies have indicated an important role for major groove contacts in Oct4-DNA recognition; however, the contributions of the RK motif in the POUh domain and the linker segment joining the two DNA-binding domains remain poorly understood. Here, by combining molecular modelling and functional assays, we find that the RK motif is essential for Oct4-DNA association by recognizing the narrowed DNA minor groove. Intriguingly, computational simulations reveal that the function of the RK motif may be finely tuned by H-bond interactions with the partially disordered linker segment and that breaking these interactions significantly enhances the DNA binding and reprogramming activities of Oct4. These findings uncover a self-regulatory mechanism for specific Oct4-DNA recognition and provide insights into the functional crosstalk at the molecular level that may illuminate mechanistic studies of the Oct protein family and possibly transcription factors in the POU family. Our gain-of-function Oct4 mutants might also be useful tools for use in reprogramming and regenerative medicine.