EMAIL THIS PAGE TO A FRIEND
Chemistry, an Asian journal
Surfactant-free synthesis of plasmonic tungsten oxide nanowires with visible-light-enhanced hydrogen generation from ammonia borane.
PMID 25873477
Abstract
WO3-x nanowires were successfully synthesized through a simple surfactant-free solvothermal method. These nanowires exhibit strong plasmonic absorption in the visible and near-infrared region owing to the abundant oxygen vacancies. The plasmon excitation of these WO3-x nanowires provide five times enhancement on the hydrogen generation from ammonia borane.