EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
Distribution and fate of synthetic musks in the Songhua River, Northeastern China: influence of environmental variables.
PMID 25874412
Abstract
Contamination levels and spatial and temporal distributions of six typical synthetic musks (SMs) in water and sediment of the Songhua River in Northeastern China were investigated. Experimental data for 72 water and 52 sediment samples collected at 29 sampling sites over 12 months spanning 2011-2012 showed that the Songhua River had been contaminated to different degrees at various sites separately from the river's source. The polycyclic musks 1,3,4,6,7,8-hexahydro-4,6,6,7,8,8-hexamethylcyclopenta-(g)-2-benzopyran (HHCB) (Galaxolide) and 7-acetyl-1,1,3,4,4,6-hexamethyl-1,2,3,4-tetrahydronaphthalene (AHTN) (Tonalide) were found most frequently and at the highest levels. Concentrations of HHCB were <2-37 ng/L in water and <0.5-17.5 ng/g dry weight (dw) in sediment. AHTN was <1-8 ng/L in water and <0.5-5.7 ng/g dw in sediment. Statistical relationships between SM concentrations and four environmental variables (temperature, illumination, runoff, and population density) in the Songhua River Basin were formulated. Concentration levels varied proportionately with the size of the city along the river, while the distribution patterns showed clear seasonal variations. HHCB/AHTN ratios mirrored the transfer and transmitting process of SMs. Concentrations of target compounds were correlated with each other, suggesting similar exposure sources. Environmental risk assessment of SMs presented seasonal variations and provided baseline information on SM exposure in the Songhua River Basin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
322377
Hexamethylbenzene, 99%
C12H18
429082
Hexamethylbenzene, purified by sublimation, ≥99%
C12H18