EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Feasibility of asymmetric flow field-flow fractionation coupled to ICP-MS for the characterization of wear metal particles and metalloproteins in biofluids from hip replacement patients.
PMID 25874977
Abstract
Hip replacements are used to improve the quality of life of people with orthopaedic conditions, but the use of metal-on-metal (MoM) arthroplasty has led to poor outcomes for some patients. These problems are related to the generation of micro- to nanosized metal wear particles containing Cr, Co or other elements, but the current analytical methods used to investigate the processes involved do not provide sufficient information to understand the size or composition of the wear particles generated in vivo. In this qualitative feasibility study, asymmetric flow field-flow fractionation (AF(4)) coupled with inductively coupled plasma mass spectrometry (ICP-MS) was used to investigate metal protein binding and the size and composition of wear metal particles present in serum and hip aspirates from MoM hip replacement patients. A well-established HPLC anion exchange chromatography (AEC) separation system coupled to ICP-MS was used to confirm the metal-protein associations in the serum samples. Off-line single particle ICP-MS (spICP-MS) analysis was used to confirm the approximate size distribution indicated by AF(4) of the wear particles in hip aspirates. In the serum samples, AF(4) -ICP-MS suggested that Cr was associated with transferrin (Tf) and Co with albumin (Alb) and an unidentified species; AEC-ICP-MS confirmed these associations and also indicated an association of Cr with Alb. In the hip aspirate sample, AF(4)-ICP-MS suggested that Cr was associated with Alb and Tf and that Co was associated with Alb and two unidentified compounds; AEC analysis confirmed the Cr results and the association of Co with Alb and a second compound. Enzymatic digestion of the hip aspirate sample, followed by separation using AF(4) with detection by UV absorption (280 nm), multi-angle light scattering and ICP-MS, suggested that the sizes of the Cr-, Co- and Mo-containing wear particles in a hip aspirate sample were in the range 40-150 nm. Off-line spICP-MS was used to confirm these findings for the Co- and Cr-containing nanoparticles. Whilst limited in scope, the results are sufficient to show the interaction of ions with transport proteins and give an indication of particle size, providing useful pathological indices. As such, the methods indicate a new way forward for in vivo investigation of the processes which lead to tissue necrosis and hip loosening in patients with MoM hip replacements.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A7455
Ammonium nitrate, BioXtra, ≥99.5%
H4N2O3
256064
Ammonium nitrate, 99.999% trace metals basis
H4N2O3
A3795
Ammonium nitrate, suitable for plant cell culture
H4N2O3
203343
Germanium, chips, 99.999% trace metals basis
Ge
203351
Germanium, powder, −100 mesh, ≥99.99% trace metals basis
Ge
263230
Germanium, chips, 99.999% trace metals basis
Ge
327395
Germanium, powder, −100 mesh, ≥99.999% trace metals basis
Ge
GF45727241
Germanium, sheet, 10x10mm, thickness 0.6mm, single crystal, -111, 100%
Ge
GF09629859
Germanium, sheet, 10x10mm, thickness 0.5mm, single crystal, -111, 100%
Ge
GF90754858
Germanium, disks, 20mm, thickness 1.0mm, single crystal, 100%
Ge
GF58081451
Germanium, sheet, 50x50mm, thickness 3.0mm, polycrystalline, 99.999%
Ge
GF44356978
Germanium, sheet, 25x25mm, thickness 1.0mm, single crystal, 99.999%
Ge
GF57527219
Germanium, rod, 25mm, diameter 2.0mm, polycrystalline, n-type, 99.999%
Ge
GF44621580
Germanium, sheet, 25x25mm, thickness 1.0mm, polycrystalline, 99.999%
Ge
GF73619107
Germanium, sheet, 50x50mm, thickness 0.5mm, single crystal, 99.999%
Ge
GF77697680
Germanium, microfoil, 25x25mm, thinness 0.25μm, specific density 166.3μg/cm2, 6 micron aluminum permanent support, 100%
Ge
GF04605780
Germanium, rod, 6mm, diameter 6.0mm, single crystal, 100%
Ge
GF31896366
Germanium, sheet, 7x24mm, thickness 1.0mm, polycrystalline, 99.999%
Ge
GF99720647
Germanium, sheet, 50x50mm, thickness 1.0mm, polycrystalline, 99.999%
Ge
GF46650090
Germanium, disks, 15mm, thickness 1.0mm, polycrystalline, 100%
Ge
GF58880739
Germanium, microfoil, 25x25mm, thinness 0.5μm, specific density 333μg/cm2, 6 micron aluminum permanent support, 100%
Ge
GF12210624
Germanium, microfoil, 50x50mm, thinness 0.25μm, specific density 166.3μg/cm2, 6 micron aluminum permanent support, 100%
Ge
GF65029417
Germanium, rod, 25mm, diameter 5mm, polycrystalline, n-type, 99.999%
Ge
GF14446346
Germanium, rod, 50mm, diameter 5mm, polycrystalline, n-type, 99.999%
Ge
GF09210417
Germanium, sheet, 10x10mm, thickness 0.10mm, polycrystalline, 99.999%
Ge
GF00633664
Germanium, sheet, 10x10mm, thickness 0.25mm, polycrystalline, 99.999%
Ge
GF20558022
Germanium, sheet, 25x25mm, thickness 3.0mm, polycrystalline, 99.999%
Ge
GF49319815
Germanium, sheet, 4x4mm, thickness 0.05mm, polycrystalline, 99.999%
Ge
GF37208369
Germanium, sheet, 6x6mm, thickness 1.0mm, polycrystalline, 99.999%
Ge