PloS one
Functional characterization of the vitamin K2 biosynthetic enzyme UBIAD1.
PMID 25874989
Abstract
UbiA prenyltransferase domain-containing protein 1 (UBIAD1) plays a significant role in vitamin K2 (MK-4) synthesis. We investigated the enzymological properties of UBIAD1 using microsomal fractions from Sf9 cells expressing UBIAD1 by analysing MK-4 biosynthetic activity. With regard to UBIAD1 enzyme reaction conditions, highest MK-4 synthetic activity was demonstrated under basic conditions at a pH between 8.5 and 9.0, with a DTT ≥0.1 mM. In addition, we found that geranyl pyrophosphate and farnesyl pyrophosphate were also recognized as a side-chain source and served as a substrate for prenylation. Furthermore, lipophilic statins were found to directly inhibit the enzymatic activity of UBIAD1. We analysed the aminoacid sequences homologies across the menA and UbiA families to identify conserved structural features of UBIAD1 proteins and focused on four highly conserved domains. We prepared protein mutants deficient in the four conserved domains to evaluate enzyme activity. Because no enzyme activity was detected in the mutants deficient in the UBIAD1 conserved domains, these four domains were considered to play an essential role in enzymatic activity. We also measured enzyme activities using point mutants of the highly conserved aminoacids in these domains to elucidate their respective functions. We found that the conserved domain I is a substrate recognition site that undergoes a structural change after substrate binding. The conserved domain II is a redox domain site containing a CxxC motif. The conserved domain III is a hinge region important as a catalytic site for the UBIAD1 enzyme. The conserved domain IV is a binding site for Mg2+/isoprenyl side-chain. In this study, we provide a molecular mapping of the enzymological properties of UBIAD1.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2155000
Cholesterol, European Pharmacopoeia (EP) Reference Standard
C27H46O
PHR1533
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
C27H46O
47127-U
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
C27H46O
F6892
Farnesyl pyrophosphate ammonium salt, methanol:ammonia solution, ≥95% (TLC)
C15H37N3O7P2
G6772
Geranyl pyrophosphate ammonium salt, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)
C10H20O7P2 · 3H3N
G6025
Geranylgeranyl pyrophosphate ammonium salt, solution, ≥95% (TLC), ~1 mg/mL in methanol: NH4OH (7:3)
C20H36O7P2 · xNH3
71259
Insect GeneJuice Transfection Reagent, Proprietary liposome transfection reagent optimized for maximal transfection efficiency of Sf9 insect cells for baculovirus protein expression.
1604610
Risedronate sodium, United States Pharmacopeia (USP) Reference Standard
C7H10NNaO7P2 · 2.5H2O
SML0650 Risedronate sodium, ≥97% (HPLC)
C7H10NO7P2 · Na
S6196
Simvastatin, ≥97% (HPLC), solid
C25H38O5
PHR1438
Simvastatin, Pharmaceutical Secondary Standard; Certified Reference Material
C25H38O5
1612700
Simvastatin, United States Pharmacopeia (USP) Reference Standard
C25H38O5
38956
Simvastatin, analytical standard
C25H38O5
S0650000
Simvastatin, European Pharmacopoeia (EP) Reference Standard
C25H38O5
71726
Sodium dodecyl sulfate, for ion pair chromatography, LiChropur, ≥99.0%
C12H25NaO4S
Y0000620
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
C12H25NaO4S