EMAIL THIS PAGE TO A FRIEND
Epigenetics & Chromatin
Proteogenomics analysis reveals specific genomic orientations of distal regulatory regions composed by non-canonical histone variants.
PMID 25878728