EMAIL THIS PAGE TO A FRIEND
Journal of analytical methods in chemistry
Development of a gradient HPLC method for the simultaneous determination of sotalol and sorbate in oral liquid preparations using solid core stationary phase.
PMID 25878920
Abstract
A selective and sensitive gradient HPLC-UV method for quantification of sotalol hydrochloride and potassium sorbate in five types of oral liquid preparations was developed and fully validated. The separation of an active substance sotalol hydrochloride, potassium sorbate (antimicrobial agent), and other substances (for taste and smell correction, etc.) was performed using an Ascentis Express C18 (100 × 4.6 mm, particles 2.7 μm) solid core HPLC column. Linear gradient elution mode with a flow rate of 1.3 mL min(-1) was used, and the injection volume was 5 µL. The UV/Vis absorbance detector was set to a wavelength of 237 nm, and the column oven was conditioned at 25°C. A sodium dihydrogen phosphate dihydrate solution (pH 2.5; 17.7 mM) was used as the mobile phase buffer. The total analysis time was 4.5 min (+2.5 min for reequilibration). The method was successfully employed in a stability evaluation of the developed formulations, which are now already being used in the therapy of arrhythmias in pediatric patients; the method is also suitable for general quality control, that is, not only just for extemporaneous preparations containing the mentioned substances.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50461-U
Ascentis® Express C18 PCP HPLC Columns, 2.7 μm particle size, L × I.D. 10 cm × 4.6 mm
50462-U
Ascentis® Express C18 PCP HPLC Columns, 2.7 μm particle size, L × I.D. 15 cm × 4.6 mm
50822-U
Ascentis® Express C18, 2 μm Guard Cartridge, 2 μm particle size, L × I.D. 5 mm × 2.1 mm, pkg of 3 ea
50805-U
Ascentis® Express C18, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 2 cm × 2.1 mm
50809-U
Ascentis® Express C18, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 3 cm × 2.1 mm
50811-U
Ascentis® Express C18, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 5 cm × 2.1 mm
50812-U
Ascentis® Express C18, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 7.5 cm × 2.1 mm
50813-U
Ascentis® Express C18, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 10 cm × 2.1 mm
50814-U
Ascentis® Express C18, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 15 cm × 2.1 mm
50815-U
Ascentis® Express C18, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 3 cm × 3 mm
50816-U
Ascentis® Express C18, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 5 cm × 3 mm
50817-U
Ascentis® Express C18, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 7.5 cm × 3 mm
50819-U
Ascentis® Express C18, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 10 cm × 3 mm
50821-U
Ascentis® Express C18, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 15 cm × 3 mm
53982-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 75 μm
53985-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 100 μm
53989-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 200 μm
53992-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 300 μm
53998-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 500 μm
54219-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 75 μm
54256-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 100 μm
54261-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 200 μm
54271-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 300 μm
54273-U Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 500 μm
53501-U
Ascentis® Express C18, 2.7 Micron Guard Cartridge, 2.7 μm particle size, L × I.D. 5 mm × 2.1 mm, pkg of 3 ea
53504-U
Ascentis® Express C18, 2.7 Micron Guard Cartridge, 2.7 μm particle size, L × I.D. 5 mm × 3 mm, pkg of 3 ea
53508-U
Ascentis® Express C18, 2.7 Micron Guard Cartridge, 2.7 μm particle size, L × I.D. 5 mm × 4.6 mm, pkg of 3 ea
53799-U
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 2 cm × 2.1 mm
53802-U
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 3 cm × 2.1 mm
53804-U
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 7.5 cm × 2.1 mm