EMAIL THIS PAGE TO A FRIEND
Regulatory toxicology and pharmacology : RTP
Developmental immunotoxicity testing of 4-methyl anisole.
PMID 25882306
Abstract
The developmental immunotoxicity of 4-methyl anisole (4MA) was investigated in the rat. Four study designs were used, with either premating or post-weaning onset of exposure, continued to postnatal day 50, and with or without additional oral gavage of pups from postnatal day 10 onward. Reduced litter size (benchmark dose lower confidence limit (BMDL) 80mg/kg bw/day) was the most sensitive developmental parameter, with pup relative organ weight effects observed at similar BMDLs, in the absence of maternal toxicity. Eosinophil numbers were reduced at lower doses (BMDL 16mg/kg bw/day). KLH challenge resulted in increased IL-13 and TNF-α responses, and variably reduced IgG production (BMDL 27mg/kg bw/day). T4 levels were reduced by 11% at maximum with a BMDL of 73mg/kg bw/day. Differences between exposure cohorts were limited and were considered to be without biological significance. This study shows that 4MA induces developmental immunotoxicity at doses below those inducing developmental and general toxicity. These observations being independent of the study designs applied suggest that the post-weaning period, included in all designs, is the most relevant sensitive period for inducing 4MA mediated developmental immunotoxicity. Moreover, this study stresses the importance of including developmental immunotoxicity testing by default in regulatory toxicology.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
148091
4-Methylanisole, 99%
C8H10O
W268100
4-Methylanisole, ≥99%, FCC, FG
C8H10O