EMAIL THIS PAGE TO A FRIEND
Steroids
A convenient preparation of the side-chain lactone ring of a withanolide precursor.
PMID 2588305
Abstract
Condensation of C22 and C21 steroidal aldehydes with ethyl 4,6-dimethyl-2-oxo-2H-pyran-5-carboxylate 1 in alkaline medium, followed by decarboxylation, provides a simple route to the alpha, beta-unsaturated side-chain delta-lactone synthesis for a classical withanolide precursor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E33408
Ethyl isodehydracetate, 99%
C10H12O4