EMAIL THIS PAGE TO A FRIEND
Clinical chemistry and laboratory medicine
Faecal leukocyte esterase activity is an alternative biomarker in inflammatory bowel disease.
PMID 25883206
Abstract
Leukocyte cytosolic proteins (e.g., calprotectin) are emerging biomarkers for inflammatory bowel disease. Leukocyte aryl esterase activity has been commonly used for sensitive detection of leukocytes in human body fluids such as urine. Urine test strip results are generally reported in categories. As automated strip readers allow quantitative data to be reported, sensitive quantitative detection of leukocytes in body fluids has become possible. Here, we explored the use of leukocyte esterase as a potential alternative faecal biomarker for inflammatory bowel disease. We evaluated leukocyte esterase activity in faecal extracts and compared Cobas u 411 (Roche) quantitative reflectance data with calprotectin concentration for 107 routine samples. Stability of leukocyte esterase for trypsin digestion was carried out by adding trypsin to the extract. Incubation occurred at 37 °C for 24 h or 48 h. Reproducibility of the reflectance signal was good (within-run imprecision: 6.1%; between-run imprecision: 6.2%). Results were linear in the range 103-106 WBC/100 mg faeces. The lower limit of detection was 4 WBC/μL and the lower limit of quantification was 5 WBC/μL. Stability of LE activity in stool and faecal matrix was good. An adequate correlation was obtained between leukocyte esterase activity and the faecal calprotectin concentration: log(y)=4.28+0.29log(x). In vitro experiments monitored the digestion of leukocyte esterase and faecal calprotectin. Leukocyte esterase activity was significantly less affected by trypsin activity than calprotectin immunoreactivity. Quantitative leukocyte esterase activity of faecal extracts provides information about the leukocyte count in the gut lumen. Leukocyte esterase is a promising and affordable alternative biomarker for monitoring inflammatory bowel disease.