EMAIL THIS PAGE TO A FRIEND
Nanoscale Research Letters
Self-alignment of silver nanoparticles in highly ordered 2D arrays.
PMID 25883540
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
360066
2,2,4-Trimethylpentane, anhydrous, 99.8%
C8H18
650439
2,2,4-Trimethylpentane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.5%
C8H18
34862
2,2,4-Trimethylpentane, for HPLC, ≥99%
C8H18
360597
2,2,4-Trimethylpentane, ACS reagent, ≥99.0%
C8H18
59030
2,2,4-Trimethylpentane, analytical standard
C8H18
59045
2,2,4-Trimethylpentane, puriss. p.a., ACS reagent, ≥99.5% (GC)
C8H18
32291
2,2,4-Trimethylpentane, puriss. p.a., ≥99.5% (GC)
C8H18
15-2200
2,2,4-Trimethylpentane, SAJ first grade, ≥98.0%
C8H18
15-2190
2,2,4-Trimethylpentane, JIS special grade
C8H18
15-2211
2,2,4-Trimethylpentane, for HPLC
C8H18
258776
2,2,4-Trimethylpentane, ReagentPlus®, ≥99%
C8H18
44964
Density Standard 692 kg/m3, H&D Fitzgerald Ltd. Quality
C8H18
323586
Dioctyl sulfosuccinate sodium salt, ≥97%
C20H37NaO7S
D1685
Docusate sodium, meets USP testing specifications
C20H37NaO7S
Y0001567
Docusate sodium, European Pharmacopoeia (EP) Reference Standard
C20H37NaO7S
86139
Docusate sodium salt, BioUltra, ≥99.0% (TLC)
C20H37NaO7S
D4422
Docusate sodium salt, BioXtra, ≥99%
C20H37NaO7S
86140
Docusate sodium salt, purum, ≥96.0% (TLC)
C20H37NaO7S
324558
Ethylene glycol, anhydrous, 99.8%
C2H6O2
03747
Ethylene glycol, BioUltra, ≥99.5% (GC)
C2H6O2
85978
Ethylene glycol, analytical standard
C2H6O2
1265515
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
C2H6O2
PHR1046
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
C2H6O2
24204
Ethylene glycol, puriss., ≥99.5% (GC)
C2H6O2
61941 Ethylene glycol 5 M solution
C2H6O2
721980 Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.
C2H6O2
721972 Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
C2H6O2
S8157
Silver nitrate, BioXtra, >99% (titration)
AgNO3
S7276
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
AgNO3
204390
Silver nitrate, 99.9999% trace metals basis
AgNO3