EMAIL THIS PAGE TO A FRIEND
Molecular brain
The production of viral vectors designed to express large and difficult to express transgenes within neurons.
PMID 25887710
Abstract
Viral vectors are frequently used to deliver and direct expression of transgenes in a spatially and temporally restricted manner within the nervous system of numerous model organisms. Despite the common use of viral vectors to direct ectopic expression of transgenes within the nervous system, creating high titer viral vectors that are capable of expressing very large transgenes or difficult to express transgenes imposes unique challenges. Here we describe the development of adeno-associated viruses (AAV) and lentiviruses designed to express the large and difficult to express GluN2A or GluN2B subunits of the N-methyl-D-aspartate receptor (NMDA) receptor, specifically within neurons. We created a number of custom designed AAV and lentiviral vectors that were optimized for large transgenes, by minimizing DNA sequences that were not essential, utilizing short promoter sequences of 8 widely used promoters (RSV, EFS, TRE3G, 0.4αCaMKII, 1.3αCaMKII, 0.5Synapsin, 1.1Synapsin and CMV) and utilizing a very short (~75 bps) 3' untranslated sequence. Not surprisingly these promoters differed in their ability to express the GluN2 subunits, however surprisingly we found that the neuron specific synapsin and αCaMKII, promoters were incapable of conferring detectable expression of full length GluN2 subunits and detectable expression could only be achieved from these promoters if the transgene included an intron or if the GluN2 subunit transgenes were truncated to only include the coding regions of the GluN2 transmembrane domains. We determined that viral packaging limit, transgene promoter and the presence of an intron within the transgene were all important factors that contributed to being able to successfully develop viral vectors designed to deliver and express GluN2 transgenes in a neuron specific manner. Because these vectors have been optimized to accommodate large open reading frames and in some cases contain an intron to facilitate expression of difficult to express transgenes, these viral vectors likely could be useful for delivering and expressing many large or difficult to express transgenes in a neuron specific manner.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32883
4-tert-Octylphenol monoethoxylate solution, 1 μg/mL in acetone, analytical standard
C16H26O2
32882
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
C16H26O2
MAB377
Anti-NeuN Antibody, clone A60, Anti-NeuN Antibody, clone A60 detects level of NeuN and has been published and validated for use in FC, IC, IF, IH, IH(P), IP and WB.
AP192F
Donkey Anti-Mouse IgG Antibody, FITC conjugate, Species Adsorbed, Detect Donkey Mouse IgG using this Donkey anti-Mouse IgG Antibody, FITC conjugate, Species Adsorbed validated for use in IF.
F7250
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
C21H11NO5S
47673
Formaldehyde solution, tested according to Ph.Eur.
CH2O
71383
Sodium chloride, tested according to Ph.Eur.
ClNa
71387
Sodium chloride, certified reference material for titrimetry, certified by BAM, according to ISO 17025, ≥99.5%
ClNa
PHR1321
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
ClNa
15990
Sodium pyruvate, purum, ≥99.0% (NT)
C3H3NaO3
71177
Sodium standard for AAS, ready-to-use, traceable to BAM, in H2O
ClNa
84100
Sucrose, for microbiology, ACS reagent, ≥99.0%
C12H22O11
S1600000
Sucrose, European Pharmacopoeia (EP) Reference Standard
C12H22O11
1623637
Sucrose, United States Pharmacopeia (USP) Reference Standard
C12H22O11
PHR1001
Sucrose
C12H22O11
S1174
Sucrose, analytical standard, for enzymatic assay kit SCA20
C12H22O11