EMAIL THIS PAGE TO A FRIEND
Trials
The acute effect of beta-guanidinopropionic acid versus creatine or placebo in healthy men (ABC Trial): study protocol for a randomized controlled trial.
PMID 25888414
Abstract
Despite adequate treatment, up to 30% of treated antihypertensive patients with primary, uncomplicated hypertension remain uncontrolled. We proposed that high intracellular activity of the ATP regenerating enzyme creatine kinase (CK) increases pressor responses and hypertension risk. In line with this, we found that plasma CK activity after rest, a surrogate measure of tissue activity, is the main predictor of blood pressure levels and failure of antihypertensive therapy in the general population. In addition, the creatine analog and competitive oral creatine kinase inhibitor beta-guanidinopropionic acid effectively and safely reduced blood pressure in the spontaneously hypertensive rat. However, to our knowledge there are no human data on the safety of oral supplementation with this substance. Therefore, we will assess the tolerability of beta-guanidinopropionic acid in men, compared to creatine and placebo. This is a randomized, active and placebo controlled, triple blind, double dummy, single center clinical intervention trial in 24 healthy male volunteers, 18 to 50xa0years old, recruited in the Netherlands. The intervention consists of one week of daily oral administration of beta-guanidinopropionic acid 100xa0mg, creatine 5 gram, or placebo. The primary outcome is the tolerability of beta-guanidinopropionic acid as a descriptive measure, in an intent-to-treat analysis. Other outcomes include the placebo-adjusted differences with baseline in biochemical and hemodynamic parameters, including plasma markers of muscle tissue damage, urine sodium excretion, resting sitting systolic and diastolic brachial blood pressure, supine systolic and diastolic central blood pressure, pulse wave velocity and augmentation index, heart rate, cardiac contractility, cardiac output, and total peripheral resistance. There is an unfulfilled need for new conservative options to treat resistant hypertension. This study will provide first-in-men data on creatine kinase inhibition as a potential new class of antihypertensive drugs. The Netherlands National Trial Register Trialregister.nl (identifier NTR 4444) , registered 9 March 2014.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0780
Creatine, anhydrous
C4H9N3O2