EMAIL THIS PAGE TO A FRIEND
BMC Veterinary Research
Effects of sodium citrate and acid citrate dextrose solutions on cell counts and growth factor release from equine pure-platelet rich plasma and pure-platelet rich gel.
PMID 25889052