EMAIL THIS PAGE TO A FRIEND
Clinical immunology (Orlando, Fla.)
Tuberculosis specific responses following therapy for TB: Impact of HIV co-infection.
PMID 25889622
Abstract
Characterizing perturbations in the immune response to tuberculosis in HIV can develop insights into the pathogenesis of coinfection. HIV+ TB+ and TB monoinfected (TB+) subjects recruited from clinics in Bamako prior to initiation of TB treatment were evaluated at time-points following initiation of therapy. Flow cytometry assessed CD4+/CD8+ T cell subsets and activation markers CD38/HLA-DR. Antigen specific responses to TB proteins were assessed by intracellular cytokine detection and proliferation. HIV+ TB+ subjects had significantly higher markers of immune activation in the CD4+ and CD8+ T cells compared to TB+ subjects. HIV+ TB+ had lower numbers of TB-specific CD4+ T cells at baseline. Plasma IFNγ levels were similar between HIV+ TB+ and TB+ subjects. No differences were observed in in-vitro proliferative capacity to TB antigens between HIV+ TB+ and TB+ subjects. Subjects with HIV+ TB+ coinfection demonstrate in vivo expansion of TB-specific CD4+ T cells. Immunodeficiency associated with CD4+ T cell depletion may be less significant compared to immunosuppression associated with HIV viremia or untreated TB infection.