EMAIL THIS PAGE TO A FRIEND
Journal of Translational Medicine
Identification of stroke-associated-antigens via screening of recombinant proteins from the human expression cDNA library (SEREX).
PMID 25890248