EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Preparation and evaluation of surface-bonded tricationic ionic liquid silica as stationary phases for high-performance liquid chromatography.
PMID 25890438
Abstract
Two tricationic ionic liquids were prepared and then bonded onto the surface of supporting silica materials through "thiol-ene" click chemistry as new stationary phases for high-performance liquid chromatography. The obtained columns of tricationic ionic liquids were evaluated respectively in the reversed-phase liquid chromatography (RPLC) mode and hydrophilic interaction liquid chromatography (HILIC) mode, and possess ideal column efficiency of 80,000 plates/m in the RPLC mode with naphthalene as the test solute. The tricationic ionic liquid stationary phases exhibit good hydrophobic and shape selectivity to hydrophobic compounds, and RPLC retention behavior with multiple interactions. In the HILIC mode, the retention and selectivity were evaluated through the efficient separation of nucleosides and bases as well as flavonoids, and the typical HILIC retention behavior was demonstrated by investigating retention changes of hydrophilic solutes with water volume fraction in mobile phase. The results show that the tricationic ionic liquid columns possess great prospect for applications in analysis of hydrophobic and hydrophilic samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N5893
(±)-Naringenin, ≥95%
C15H12O5
657336
1,3,5-Tris(bromomethyl)benzene, 97%
C9H9Br3
235466
1-Vinylimidazole, ≥99%
C5H6N2
441090
2,2′-Azobis(2-methylpropionitrile), 98%
C8H12N4
755745
2,2′-Azobis(2-methylpropionitrile), recrystallized from methanol, 99%
C8H12N4
516961
Ammonium formate, ≥99.995% trace metals basis
CH5NO2
78314
Ammonium formate solution, BioUltra, 10 M in H2O
CH5NO2
714690
Ammonium formate solution, 10 mM in H2O, for HPLC
CH5NO2
10798
Apigenin, ≥95.0% (HPLC)
C15H10O5
544094
Bis(trifluoromethane)sulfonimide lithium salt, 99.95% trace metals basis
C2F6LiNO4S2
449504
Bis(trifluoromethane)sulfonimide lithium salt
C2F6LiNO4S2
15224
Bis(trifluoromethane)sulfonimide lithium salt, puriss., ≥99.0% (19F-NMR)
C2F6LiNO4S2
C80105
Chrysin, 97%
C15H10O4
D7802
Daidzein, ≥98%, synthetic
C15H10O4
30408
Daidzin, ≥95.0% (HPLC)
C21H20O9
47752
Formononetin, ≥99.0% (TLC)
C16H12O4
G6649
Genistein, synthetic, ≥98% (HPLC), powder
C15H10O5
G6776
Genistein, from Glycine max (soybean), ~98% (HPLC)
C15H10O5
W431300
Hesperetin, ≥95%
C16H14O6
H5254
Hesperidin, ≥80%
C28H34O15
W530098
Naringenin, natural (US), 98%
C15H12O5
71162
Naringin, ≥95% (HPLC)
C27H32O14
N1376
Naringin, ≥90%, from citrus fruit
C27H32O14
P1659
Pelargonidin chloride
C15H11ClO5
P7912
Phloretin, ≥99%
C15H14O5
82415
Prunetin, ≥98.0% (TLC)
C16H12O5