EMAIL THIS PAGE TO A FRIEND
American journal of surgery
Diclofenac causes anastomotic leakage in the proximal colon but not in the distal colon of the rat.
PMID 25890814
Abstract
Nonsteroidal anti-inflammatory drugs have been associated with anastomotic leakage. It was studied if diclofenac affects anastomoses differently depending on the location in the gut. Ninety-five rats were randomized to 6 groups with an anastomosis in either ileum (IL), proximal colon (PC), or distal colon (DC). Groups IL+ (n = 10), PC+ (n = 30), and DC+ (n = 10) received diclofenac (3 mg/kg/day) from day 0 until sacrifice on day 3. Group PC- (n = 15) did not receive diclofenac. Groups PC1+ and PC2+ (n = 15 each) were given diclofenac from day 1 to 4 and from day 2 to 5. Leak rates were 10/10 in group IL+, 22/30 in PC+, 1/10 in DC+, and 1/15 in PC-. Delayed administration of diclofenac by 1 or 2 days (6/15, P = .05) resulted in reduced leakage rates. Mechanical strength results corresponded with leak rates. Diclofenac causes leakage of anastomoses in rat IL and PC, but not in the DC. This suggests a role for the ileal and proximal colonic content in diclofenac-induced leakage.