EMAIL THIS PAGE TO A FRIEND
Toxicology in vitro : an international journal published in association with BIBRA
Cell compatibility of a maghemite/polymer biomedical nanoplatform.
PMID 25891827
Abstract
We are reporting the cytocompatibility and cellular fate of an iron oxide/polymer nanoplatform (IONP) in its most basic formulation, using both mesenchymal (vascular smooth muscle cells, VSMC), and epithelial (opossum kidney, OK) cells. The cytotoxicity and cell internalization of the nanoplatform has been evaluated in relation to time of exposure and concentration of different components. A series of samples with different iron oxide nanoparticle, sizes, hydrodynamic sizes and iron/polymer ratio have been examined. In all cases cytotoxicity is low, and it is mostly determined by the internalization rate, being higher in VSMC than in OK cells. The mean lethal dose has a very narrow threshold, and necrosis is the only cell death type. IONP uptake shows little incidence on oxidative stress, and inflammasome activation is only observed with the smaller IONP at high concentration. The internalization rate in VSMC is determined by the polymer concentration exclusively. In OK cells, internalization rate seems to increase with decreasing hydrodynamic size. Internalization occurs through clathrin-dependent endocytosis, as it is prevented by potassium depletion and chlorpromazine. IONP are directed and accumulated in lysosomes. Under IONP overload, lysosomal dysfunction would cause cell death using concentrations that are hardly achieved in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D2773
Dithranol, ≥90% (HPLC)
C14H10O3
132101
Sodium trifluoroacetate, 98%
C2F3NaO2