PLoS ONE
MKAN27435 is required for the biosynthesis of higher subclasses of lipooligosaccharides in Mycobacterium kansasii.
PMID 25893968
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
191760
3,4-Diaminobenzophenone, 97%
C13H12N2O
F2252
L-(−)-Fucose, ≥99%
C6H12O5