EMAIL THIS PAGE TO A FRIEND
Journal of clinical hypertension (Greenwich, Conn.)
Plasma Homocysteine is a Predictive Factor for Arterial Stiffness: A Community-Based 4.8-Year Prospective Study.
PMID 25894449
Abstract
The authors investigated whether plasma total homocysteine (tHcy) is a predictive factor for arterial stiffness (carotid-femoral pulse wave velocity [cf-PWV] and carotid-radial PWV) in 1447 patients from a 4.8-year prospective study in Beijing, People's Republic of China. Baseline tHcy showed a significant relationship with follow-up cf-PWV (β=0.817, P=.015) in a multivariable linear regression analysis. A stepwise logistic regression model showed that baseline levels of tHcy were significantly associated with follow-up cf-PWV in the adjusted models. Furthermore, the baseline tHcy levels showed a significant association with increases in cf-PWV. There was no association between the change in tHcy and increase in PWV. The present study clearly demonstrated an association between tHcy levels and arterial stiffness, indicating that tHcy is an independent predictive factor for arterial stiffness in a community-based population.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H4628
DL-Homocysteine, ≥95% (titration)
C4H9NO2S