EMAIL THIS PAGE TO A FRIEND
Applied biochemistry and biotechnology
Bioconversion of Birch Wood Hemicellulose Hydrolyzate to Xylitol.
PMID 25894947
Abstract
A sugar solution containing 42.9 g l(-1) of xylose was prepared from the wood of Japanese white birch (Betula platyphylla var. japonica) by hydrolysis with 3 % sulfuric acid with a liquor-to-solid ratio of 4 (g g(-1)) at 120 °C for 1 h. During the acid hydrolysis, undesirable by-products were generated, such as acetic acid, furfural, and low-molecular-weight phenols, which inhibit bioconversion of xylose to xylitol. These inhibitors were successfully removed from the hydrolyzate by sorption onto a steam-activated charcoal followed by treatment with an anion exchange resin. Bioconversion of the detoxified hydrolyzate to xylitol by the yeast Candida magnoliae was investigated under the microaerobic conditions. The oxygen transfer rate (OTR) varied from 9.6 to 22.3 mmol O2 l(-1) h(-1). The best fermentative performance of C. magnoliae in the birch wood hydrolyzate (xylitol yield 0.74 g xylitol g xylose(-1); volumetric productivity 1.0 g l(-1) h(-1)) was obtained at the OTR of 12.6 mmol O2 l(-1) h(-1).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
X3375
Xylitol, ≥99%
C5H12O5
W507930
Xylitol
C5H12O5