EMAIL THIS PAGE TO A FRIEND
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging
Characterization of [(99m)Tc]Duramycin as a SPECT Imaging Agent for Early Assessment of Tumor Apoptosis.
PMID 25896815
Abstract
We investigated the usefulness of [(99m)Tc]duramycin for monitoring early response to cancer therapy in mice, with an eye towards clinical translation. [(99m)Tc]Duramycin was injected in healthy CD1-/- mice to estimate human [(99m)Tc]duramycin radiation dose. [(99m)Tc]Duramycin single-photon emission computed tomography (SPECT) imaging of apoptosis was evaluated in a mouse model of colorectal cancer treated with irinotecan and validated ex vivo using autoradiography, cleaved caspase-3, and TdT-mediated dUTP nick-end labeling (TUNEL) histology of the tumors. The mean effective dose was estimated to be 3.74 × 10(-3) ± 3.43 × 10(-4) mSv/MBq for non-purified and 3.19 × 10(-3) ± 2.16 × 10(-4) mSv/MBq for purified [(99m)Tc]duramycin. [(99m)Tc]Duramycin uptake in vivo following therapy increased significantly in apoptotic irinotecan-treated tumors (p = 0.008). Radioactivity in the tumors positively correlated with cleaved caspase-3 (r = 0.85, p < 0.001) and TUNEL (r = 0.92, p < 0.001) staining. [(99m)Tc]Duramycin can be used to detect early chemotherapy-induced tumor cell death, and thus, may be a prospective candidate for clinical SPECT imaging of tumor response to therapy.