EMAIL THIS PAGE TO A FRIEND
Nature communications
Dynamic caveolae exclude bulk membrane proteins and are required for sorting of excess glycosphingolipids.
PMID 25897946
Abstract
Caveolae have long been implicated in endocytosis. Recent data question this link, and in the absence of specific cargoes the potential cellular function of caveolar endocytosis remains unclear. Here we develop new tools, including doubly genome-edited cell lines, to assay the subcellular dynamics of caveolae using tagged proteins expressed at endogenous levels. We find that around 5% of the cellular pool of caveolae is present on dynamic endosomes, and is delivered to endosomes in a clathrin-independent manner. Furthermore, we show that caveolae are indeed likely to bud directly from the plasma membrane. Using a genetically encoded tag for electron microscopy and ratiometric light microscopy, we go on to show that bulk membrane proteins are depleted within caveolae. Although caveolae are likely to account for only a small proportion of total endocytosis, cells lacking caveolae show fundamentally altered patterns of membrane traffic when loaded with excess glycosphingolipid. Altogether, these observations support the hypothesis that caveolar endocytosis is specialized for transport of membrane lipid.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6499 CTB, ≥98% (HPLC)
C16H13ClF3NO2
O9381
Okadaic acid from Prorocentrum concavum, 92-100% (HPLC)
C44H68O13