EMAIL THIS PAGE TO A FRIEND
Annals of nuclear medicine
[(18)F]-FLT PET to predict early response to neoadjuvant therapy in KRAS wild-type rectal cancer: a pilot study.
PMID 25899481
Abstract
This pilot study evaluated the utility of 3'-deoxy-3'[18F]-fluorothymidine ([(18)F]-FLT) positron emission tomography (PET) to predict response to neoadjuvant therapy that included cetuximab in patients with wild-type KRAS rectal cancers. Baseline [(18)F]-FLT PET was collected prior to treatment initiation. Follow-up [(18)F]-FLT was collected after three weekly infusions of cetuximab, and following a combined regimen of cetuximab, 5-FU, and radiation. Imaging-matched biopsies were collected with each PET study. Diminished [(18)F]-FLT PET was observed in 3/4 of patients following cetuximab treatment alone and in all patients following combination therapy. Reduced [(18)F]-FLT PET following combination therapy predicted disease-free status at surgery. Overall, [(18)F]-FLT PET agreed with Ki67 immunoreactivity from biopsy samples and surgically resected tissue, and was predictive of treatment-induced rise in p27 levels. These results suggest that [(18)F]-FLT PET is a promising imaging biomarker to predict response to neoadjuvant therapy that included EGFR blockade with cetuximab in patients with rectal cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
323616
3′-Deoxythymidine, 98%
C10H14N2O4