EMAIL THIS PAGE TO A FRIEND
Luminescence : the journal of biological and chemical luminescence
Micellar effect on the photophysics of heteroleptic ruthenium(II)-phenanthrolinedisulfonato complexes.
PMID 25900090
Abstract
Luminescent heteroleptic ruthenium(II) complexes of type RuLn X(3-n) [L = 1,10-phenanthroline (phen), X = 4,7 diphenyl phenanthroline disulfonate, (dpsphen) n = 0,1,2,3] were synthesized and their photophysical properties investigated in homogeneous and cationic (CTAB), anionic (SDS) and nonionic (Triton X-100) micelles. The luminescent quantum yield and lifetime of the complexes were found to increase in the presence of micellar media and on the introduction of a disulfonate ligand into the coordination sphere. Both electrostatic and hydrophobic interactions play an important role in the micellar media. Thus, by changing the nature of the ligands and the medium, we were able to tune the photophysical properties of Ru(II) complexes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
131377
1,10-Phenanthroline, ≥99%
C12H8N2