EMAIL THIS PAGE TO A FRIEND
Journal of AOAC International
Bioassay-Guided Chromatographic Isolation and Identification of Antibacterial Compounds from Artemisia annua L. That Inhibit Clostridium perfringens Growth.
PMID 25902977
Abstract
Clostridium perfringens is the causative agent of necrotic enteritis leading to significant losses in the poultry industry. Dichloromethane and n-hexane extracts of aerial parts of Artemisia annua (Asteraceae) exhibited activity against C. perfringens with minimum inhibitory concentrations (MIC) of 185 and 270 μg/mL, respectively. Bioassay-guided fractionation of the extracts gave several active fractions (MIC between 75 and 600 μg/mL). Investigations of the most active fractions resulted in the isolation and characterization of the polyacetylene ponticaepoxide (MIC between 100 and 200 μg/mL) and (+)-threo-(5E)-trideca-1,5-dien-7,9,11-triyne-3,4-diol (MIC between 400 and 800 μg/mL), the flavonols chrysosplenol D (MIC between 200 and 400 μg/mL) and casticin (slight inhibition at 800 μg/mL), and 2,4-dihydroxy-6-methoxyacetophenone (slight inhibition at 800 μg/mL). Also, the coumarin scopoletin and the sesquiterpene lactone artemisinin were isolated from active fractions but showed no inhibition of C. perfringens growth at 800 and 2000 μg/mL, respectively. Fractions containing essential oil components with camphor constituting >60% did not show inhibition of C. perfringens up to 1600 μg/mL. Extracts and some active fractions showed higher antibacterial effect than individual bioactive compounds, suggesting that synergistic effects may underlie the observed antibacterial effect. The present study adds new valuable information on the antibacterial effect of A. annua against C. perfringens.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4261
Coumarin, ≥99% (HPLC)
C9H6O2
W526509
Coumarin, ≥98%
C9H6O2
72609
Coumarin, certified reference material, TraceCERT®
C9H6O2
01260595
Coumarin, primary reference standard
C9H6O2
Y0000438
Coumarin, European Pharmacopoeia (EP) Reference Standard
C9H6O2
151947
Methanol-d4, ≥99.8 atom % D
CD4O
441384
Methanol-d4, ≥99.8 atom % D
CD4O
343803
Methanol-d4, ≥99.8 atom % D, contains 0.03 % (v/v) TMS
CD4O
611646
Methanol-d4, ≥99.8 atom % D, contains 0.05 % (v/v) TMS
CD4O
422878
Methanol-d4, 99 atom % D
CD4O
444758
Methanol-d4, "100%", 99.96 atom % D
CD4O
569534
Methanol-d4, anhydrous, ≥99.8 atom % D
CD4O
439029
Methanol-d4, ≥99.8 atom % D, contains 0.1 % (v/v) TMS
CD4O
417653
Methanol-d4, ≥99.8 atom % D, contains 1 % (v/v) TMS
CD4O
721948 Methanol-d4, NMR reference standard, 99.8 atom % D, NMR tube size 5 mm × 8 in.
CD4O
721921 Methanol-d4, NMR reference standard, 99.8 atom % D, NMR tube size 3 mm × 8 in.
CD4O
535435
Methanol-d4, "100%", ≥99.96 atom % D, contains 0.03 % (v/v) TMS
CD4O
77634
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
C8H8O3
87921
Tetramethylsilane, ≥99.0% (GC)
C4H12Si
T24007
Tetramethylsilane, ACS reagent, NMR grade, ≥99.9%
C4H12Si
87920
Tetramethylsilane, analytical standard, for NMR spectroscopy, ACS reagent
C4H12Si
523771
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
C4H12Si
V1104
Vanillin, ReagentPlus®, 99%
C8H8O3
W310727
Vanillin, natural, ≥97%, FCC, FG
C8H8O3
W310700
Vanillin, ≥97%, FCC, FG
C8H8O3
PHR1245
Vanillin, Pharmaceutical Secondary Standard; Certified Reference Material
C8H8O3
94752
Vanillin, tested according to Ph. Eur.
C8H8O3
30304
Vanillin, certified reference material, TraceCERT®
C8H8O3
V0050000
Vanillin, European Pharmacopoeia (EP) Reference Standard
C8H8O3
PHR1091
Vanillin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
C8H8O3