EMAIL THIS PAGE TO A FRIEND
Journal of AOAC International
Identification and Quantitation of Volatile Organic Compounds in Poly(methyl methacrylate) Kitchen Utensils by Headspace Gas Chromatography/Mass Spectrometry.
PMID 25902999
Abstract
A headspace GC/MS method was developed for identification and quantitation of residual volatile organic compounds in poly(methyl methacrylate) (PMMA) kitchen utensils. A sample was cut into small pieces, then N,N-dimethylacetamide was added in a headspace vial and sealed. After storing for more than 1 day at room temperature, the vial was incubated for 1 h at 90°C, and the headspace gas was analyzed by GC/MS. In 24 PMMA kitchen utensils, 16 volatile organic compounds including methyl methacrylate, methyl acrylate, toluene, 2-methyl-1-butene, 2-methyl-2-butene, 2-methylpropanal, methyl propionate, methyl isobutyrate, trans-3-heptene, heptane, cis-3-heptene, trans-2-heptene, cis-2-heptene, 2,4,4-trimethyl-1-pentene, 2,4,4-trimethyl-2-pentene, and 1-octene were identified and quantitated. These 15 volatile compounds except methyl methacrylate were found for the first time in PMMA kitchen utensils. Recovery rates from spiked samples were 97.4-104.0% with CV values of 2.8-9.6%. Samples contained 190-7900 μg/g of methyl methacrylate, 26-810 μg/g of methyl acrylate, and 2-1300 μg/g of toluene; other compounds were at levels less than 100 μg/g. Methyl methacrylate was the main monomer of PMMA and methyl acrylate was a comonomer; toluene should be used as a solvent.