Glycoconjugate journal
Pectin from Prunus domestica L. induces proliferation of IEC-6 cells through the alteration of cell-surface heparan sulfate on differentiated Caco-2 cells in co-culture.
PMID 25903683
Dietary fiber intake provides various physiological and metabolic effects for human health. Pectin, a water-soluble dietary fiber, induces morphological changes of the small intestine in vivo. However, the molecular mechanisms underlying pectin-derived morphological alterations have not been elucidated. Previously, we found that pectin purified from Prunus domestica L. altered the sulfated structure of cell-surface heparan sulfate (HS) on differentiated Caco-2 cells via fibronectin and α5β1 integrin. In this study, we investigated the biological significance of the effect of pectin on HS in differentiated Caco-2 cells. An in vitro intestinal epithelium model was constructed by co-culture of differentiated Caco-2 cells and rat IEC-6 cells, which were used as models of intestinal epithelium and intestinal crypt cells, respectively. We found that pectin-treated differentiated Caco-2 cells promoted growth of IEC-6 cells. Real-time RT-PCR analysis and western blotting showed that relative mRNA and protein expression levels of Wnt3a were upregulated by pectin treatment in differentiated Caco-2 cells. Analysis by surface plasmon resonance spectroscopy demonstrated that pectin-induced structural alteration of HS markedly decreased the interaction with Wnt3a. However, depression in the secretion of Wnt3a from Caco-2 cells by anti-Wnt3a antibody did not affect the proliferation of IEC-6 cells in co-culture system. These observations indicated that pectin altered the sulfated structure of cell-surface HS to promote secretion of Wnt3a from differentiated Caco-2 cells and Wnt3a indirectly stimulated the proliferation of IEC-6 cells.