EMAIL THIS PAGE TO A FRIEND
European Journal of Mass Spectrometry (Chichester, England)
Rapid and sensitive determination of diacetylpolyamines in human fingernail by ultraperformance liquid chromatography coupled with electrospray ionization tandem mass spectrometry.
PMID 25905872