PLoS ONE
Ethylene Response Factors Are Controlled by Multiple Harvesting Stresses in Hevea brasiliensis.
PMID 25906196
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
392707
Methyl jasmonate, 95%
C13H20O3
W341002
Methyl jasmonate, ≥98%, stabilized, FG
C13H20O3