EMAIL THIS PAGE TO A FRIEND
The Journal of heart and lung transplantation : the official publication of the International Society for Heart Transplantation
The role of the endothelin-1 pathway as a biomarker for donor lung assessment in clinical ex vivo lung perfusion.
PMID 25907141
Abstract
Normothermic ex vivo lung perfusion (EVLP) is a preservation technique that allows reassessment of donor lungs before transplantation. We hypothesized that the endothelin-1 (ET-1) axis would be associated with donor lung performance during EVLP and recipient outcomes after transplantation. ET-1, Big ET-1, endothelin-converting enzyme (ECE), and nitric oxide (NO) metabolites were quantified in the perfusates of donor lungs enrolled in a clinical EVLP trial. Lungs were divided into 3 groups: (I) Control: bilateral transplantation with good early outcomes defined as absence of primary graft dysfunction (PGD) Grade 3 (PGD3) ; (II) PGD3: bilateral lung transplantation with PGD3 any time within 72 hours; and (III) Declined: lungs rejected after EVLP. There were 25 lungs in Group I, 7 in Group II, and 16 in Group III. At 1 and 4 hours of EVLP, the perfusates of Declined lungs had significantly higher levels of ET-1 (3.1 ± 2.1 vs. 1.8±2.3 pg/ml, p = 0.01; 2.7 ± 2.2 vs. 1.3 ± 1.1 pg/ml, p = 0.007) and Big ET-1 (15.8 ± 14.2 vs. 7.0 ± 6.5 pg/ml, p = 0.001; 31.7 ± 17.4 vs. 19.4 ± 9.5 pg/ml, p = 0.007) compared with Controls. Nitric oxide metabolite concentrations were significantly higher in Declined and PGD3 lungs than in Controls. For cases of donation after cardiac death, PGD3 and Declined lungs had higher ET-1 and Big ET-1 levels at 4 hours of perfusion compared with Controls. At this time point, Big ET-1 had excellent accuracy to distinguish PGD3 (96%) and Declined (92%) from Control lungs. In donation after cardiac death lungs, perfusate ET-1 and Big ET-1 are potential predictors of lung function during EVLP and after lung transplantation. They were also associated with non-use of lungs after EVLP and thus could represent useful biomarkers to improve the accuracy of donor lungs selection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E7764
Endothelin 1, ≥97% (HPLC), powder
C109H159N25O32S5