EMAIL THIS PAGE TO A FRIEND
BMC biology
Expression of leukemia inhibitory factor in Müller glia cells is regulated by a redox-dependent mRNA stability mechanism.
PMID 25907681
Abstract
Photoreceptor degeneration is a main hallmark of many blinding diseases making protection of photoreceptors crucial to prevent vision loss. Thus, regulation of endogenous neuroprotective factors may be key for cell survival and attenuation of disease progression. Important neuroprotective factors in the retina include H2O2 generated by injured photoreceptors, and leukemia inhibitory factor (LIF) expressed in Müller glia cells in response to photoreceptor damage. We present evidence that H2O2 connects to the LIF response by inducing stabilization of Lif transcripts in Müller cells. This process was independent of active gene transcription and p38 MAPK, but relied on AU-rich elements (AREs), which we identified within the highly conserved Lif 3'UTR. Affinity purification combined with quantitative mass spectrometry identified several proteins that bound to these AREs. Among those, interleukin enhancer binding factor 3 (ILF3) was confirmed to participate in the redox-dependent Lif mRNA stabilization. Additionally we show that KH-type splicing regulatory protein (KHSRP) was crucial for maintaining basal Lif expression levels in non-stressed Müller cells. Our results suggest that H2O2-induced redox signaling increases Lif transcript levels through ILF3 mediated mRNA stabilization. Generation of H2O2 by injured photoreceptors may thus enhance stability of Lif mRNA and therefore augment neuroprotective LIF signaling during degenerative conditions in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB302
Anti-Glutamine Synthetase Antibody, clone GS-6, Detect Glutamine Synthetase using this Anti-Glutamine Synthetase Antibody, clone GS-6 validated for use in ELISA, IH, IH(P) & WB with more than 45 product citations.
W302902
D-Sorbitol, ≥98%, FCC, FG
C6H14O6
240850
D-Sorbitol, 99%
C6H14O6
S1876
D-Sorbitol, ≥98%
C6H14O6
85529
D-Sorbitol, BioUltra, ≥99.5% (HPLC)
C6H14O6
S6021
D-Sorbitol, for molecular biology, ≥98%
C6H14O6
S7547
D-Sorbitol, BioXtra, ≥98%
C6H14O6
S3889
D-Sorbitol, BioReagent, cell culture tested, plant cell culture tested
C6H14O6
309532
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
C6H10O6
T0251 Thymidine 5′-triphosphate sodium salt, ≥96%
C10H16N2O14P3Na
SRP3177
TNF-α human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested
T7539
Tumor Necrosis Factor-α from mouse, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
T5944
Tumor Necrosis Factor-α from rat, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
T0157
Tumor Necrosis Factor-α human, TNF-α, recombinant, expressed in yeast, buffered aqueous solution, suitable for cell culture
T6674
Tumor Necrosis Factor-α human, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture