EMAIL THIS PAGE TO A FRIEND
The American journal of cardiology
Orbital Atherectomy for Treating De Novo Severely Calcified Coronary Narrowing (1-Year Results from the Pivotal ORBIT II Trial).
PMID 25910525
Abstract
Percutaneous coronary intervention of severely calcified lesions has historically been associated with major adverse cardiac event (MACE) rates as high as 30%. In the ORBIT II (Evaluate the Safety and Efficacy of OAS in Treating Severely Calcified Coronary Lesions) trial, treatment of de novo severely calcified lesions with the Diamondback 360° Coronary Orbital Atherectomy System (OAS) resulted in low rates of procedural and 30-day adverse ischemic events. The long-term results from this trial have not been reported. We sought to determine the 1-year outcomes after orbital atherectomy of severely calcified coronary lesions. ORBIT II was a single-arm trial enrolling 443 subjects at 49 US sites with severely calcified lesions usually excluded from randomized trials. OAS utilizes a centrifugal differential sanding mechanism of action for plaque modification prior to stent implantation. After OAS drug-eluting stents were implanted in 88.2% of the patients. The primary safety end point was 30-day MACE, the composite of cardiac death, myocardial infarction, or target vessel revascularization [TVR]. The present analysis reports the 1-year follow-up results from ORBIT II. One-year data were available in 433 of 443 patients (97.7%), with median follow-up time of 16.7 months. The 1-year MACE rate was 16.4%, including cardiac death (3.0%), myocardial infarction (9.7%), and target vessel revascularization (5.9%). The 1-year target lesion revascularization rate was 4.7%, and stent thrombosis occurred in 1 patient (0.2%). Independent predictors of 1-year MACE and target vessel revascularization were diameter stenosis at baseline and the use of bare-metal stents. In patients with severely calcified lesions who underwent percutaneous coronary intervention, the use of OAS was associated with low rates of 1-year adverse ischemic events compared with historical controls. This finding has important clinical implications for the selection of optimum treatment strategies for patients with severely calcified lesions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
774278
Diethyl sulfite, 98%
C4H10O3S
218944
Diethylstilbestrol, mixture of cis and trans, 97%
C18H20O2