EMAIL THIS PAGE TO A FRIEND
International journal of biological macromolecules
Human urinary renalase lacks the N-terminal signal peptide crucial for accommodation of its FAD cofactor.
PMID 25910647
Abstract
Renalase is a recently discovered secretory protein involved in the regulation of blood pressure. Cells synthesize all known isoforms of human renalase (1 and 2) as flavoproteins. Accommodation of FAD in the renalase protein requires the presence of its N-terminal peptide. However, in secretory proteins, such peptides are usually cleaved during their export from the cell. In the present study, we have isolated human renalase from urinary samples of healthy volunteers and human recombinant renalases 1 and 2 expressed in Escherichia coli cells. In these proteins, we investigated the presence of the renalase N-terminal peptide and the FAD cofactor and performed computer-aided molecular analysis of the renalase crystal structure to evaluate possible consequences of removal of the N-terminal peptide. In contrast to human recombinant renalase isoforms 1 and 2 containing non-covalently bound FAD and clearly detectable N-terminal peptide, renalase purified from human urine lacks both the N-terminal signal peptide and FAD. The computer-aided analysis indicates that the removal of this peptide results in inability of the truncated renalase to bind the FAD cofactor. Thus, our results indicate that human renalase secreted in urine lacks its N-terminal peptide, and therefore catalytic activities of urinary renalase reported in the literature cannot be attributed to FAD-dependent mechanisms. We suggest that FAD-dependent catalytic functions are intrinsic properties of intracellular renalases, whereas extracellular renalases act in FAD- and possibly catalytic-independent manner.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
164194
Heptafluorobutyric acid, 98%
C4HF7O2
77249
Heptafluorobutyric acid, ≥99.0% (GC)
C4HF7O2