EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Heart and circulatory physiology
Effect of docosahexaenoic acid monoacylglyceride on systemic hypertension and cardiovascular dysfunction.
PMID 25910811
Abstract
ω-3 Fatty acid supplementation has been associated with lower blood pressure. Cardiovascular diseases are also known to be linked directly to an increase in ω-6 and a reduction in ω-3 fatty acid levels in blood circulation and tissues. To determine the effect of docosahexaenoic acid monoglycerides (MAG-DHA) on blood pressure, lipid profiles, and vascular remodeling in rats fed a high-fat/high-carbohydrate (HFHC) diet. Studies were performed in male rats subjected to 8 wk of HFHC diet supplemented or not with 3 g/day MAG-DHA. After 8 wk of daily MAG-DHA treatment, rats in the HFHC + MAG-DHA group had lower arterial blood pressure and heart rate compared with the HFHC group. Moreover, MAG-DHA prevented the increase aortic wall thickness, whereas lipid analysis of aortic tissues revealed an increase in DHA/AA ratio correlated with the production of resolvin D2 and D3 metabolites. Histological analysis revealed that MAG-DHA prevented the development of LVH in the HFHC group. Serum lipid profile analysis further showed a decrease in total cholesterol (TC) and LDL, including very low-density lipoprotein (VLDL) and triglyceride (TG) levels, together with an increase in HDL levels after 8 wk of MAG-DHA treatment compared with the HFHC group. Furthermore, daily MAG-DHA treatment resulted in reduced proinflammatory marker levels such as CRP, IL-6, TNFα, and IL-1β. Altogether, these findings revealed that per os administration of MAG-DHA prevents HFHC-diet induced hypertension and LVH in rats.