EMAIL THIS PAGE TO A FRIEND
Arthroscopy : the journal of arthroscopic & related surgery : official publication of the Arthroscopy Association of North America and the International Arthroscopy Association
Local Application of Gelatin Hydrogel Sheets Impregnated With Platelet-Derived Growth Factor BB Promotes Tendon-to-Bone Healing After Rotator Cuff Repair in Rats.
PMID 25911389
Abstract
To determine whether the local application of platelet-derived growth factor BB (PDGF-BB) in hydrogel sheets would promote healing and improve histologic characteristics and biomechanical strength after rotator cuff (RC) repair in rats. To assess the effect of PDGF-BB on tendon-to-bone healing we divided 36 adult male Sprague-Dawley rats treated with bilateral surgery to repair the supraspinatus tendon at its insertion site into 3 groups: group 1 = suture-only group; group 2 = suture and gelatin hydrogel sheets impregnated with phosphate-buffered saline (PBS); and group 3 = suture and gelatin hydrogel sheets impregnated with PDGF-BB (0.5 μg). Semiquantitative histologic evaluation was carried out 2, 6, and 12 weeks later; cell proliferation was assessed 2 and 6 weeks postoperatively by immunostaining for proliferating cell nuclear antigen (PCNA), and biomechanical testing, including ultimate load to failure, stiffness, and ultimate stress to failure, was performed 12 weeks after the operation. At 2 weeks, the average percentage of PCNA-positive cells at the insertion site was significantly higher in group 3 (40.5% ± 2.4%) than in group 1 (32.1% ± 6.9%; P = .03) and group 2 (31.9% ± 3.7%; P = .02). At 2 and 6 weeks, the histologic scores were similar among the 3 groups. At 12 weeks, the histologic score was significantly higher in group 3 (10.3 ± 0.8) than in group 1 (8.5 ± 0.5; P = .002) or group 2 (8.8 ± 0.8; P = .009), whereas ultimate load to failure, stiffness, and ultimate load to stress (normal control population, 44.73 ± 9.75 N, 27.59 ± 4.32 N/mm, and 21.33 ± 4.65 N/mm(2), respectively) were significantly higher in group 3 (28.28 ± 6.28 N, 11.05 ± 2.37 N/mm, and 7.99 ± 2.13 N/mm(2), respectively) than in group 1 (10.44 ± 1.98 N, 4.74 ± 1.31 N/mm, and 3.28 ± 1.27 N/mm(2), respectively; all P < .001) or group 2 (11.85 ± 2.89 N, 5.86 ± 1.75 N/mm, and 3.31 ± 0.80 N/mm(2), respectively; all P < .001). The placement of a PDGF-BB-impregnated hydrogel sheet just lateral to a transected and acutely reattached supraspinatus tendon produced significantly more PCNA-positive cells at 2 weeks and greater collagen fiber orientation, ultimate failure loads, stiffness, and stress to failure at 12 weeks than did a PBS-impregnated hydrogel sheet. No differences in vascularity or cellularity were observed. The local application of PDGF-BB-impregnated gelatin hydrogel may help to promote tendon-to-bone healing after RC repair in humans.