EMAIL THIS PAGE TO A FRIEND
Aquatic toxicology (Amsterdam, Netherlands)
Effects of the anti-androgen cyproterone acetate (CPA) on oocyte meiotic maturation in rainbow trout (Oncorhynchus mykiss).
PMID 25911576
Abstract
In the present study, we aimed at characterizing the effect of cyproterone acetate (CPA), an anti-androgenic compound, on oocyte meiotic maturation in a freshwater teleost fish species, the rainbow trout (Oncorhynchus mykiss). Fully-grown post-vitellogenic ovarian follicles were incubated in vitro with CPA, luteinizing hormone (Lh) or a combination of CPA and Lh. Incubations were also performed using a combination of Lh and testosterone (T). The occurrence of oocyte maturation (i.e., resumption of the meiotic process) was assessed by monitoring germinal vesicle breakdown (GVBD) after a 72h in vitro incubation. The effect of CPA on the production of 17,20β-dihydroxy-4-pregnen-3-one (17,20βP), the natural maturation-inducing steroid (MIS), was quantified by radioimmunoassay. Our results show that CPA dramatically inhibits Lh-induced oocyte maturation and MIS synthesis. We also observed a synergistic effect of Lh and T on oocyte maturation in highly competent oocytes (i.e., able to resume meiosis after stimulation by low doses of Lh). Our results also show that a combination of CPA and Lh inhibits phosphorylation of extracellular signal-regulated kinase (Erk), kinases that are associated with oocyte maturation in many species. As a whole, our results indicate that CPA has a potential to alter meiotic maturation in rainbow trout. Further analyses are, however, needed to determine the mechanisms by which this anti-androgen interferes with the meiotic process. Furthermore, the present study provides a framework for better understanding of the ecological consequences of exposure to anti-androgens and resulting meiotic maturation abnormalities observed in trout.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3412 Cyproterone acetate, ≥98%
C24H29ClO4
F9397
Flutamide
C11H11F3N2O3