EMAIL THIS PAGE TO A FRIEND
Journal of cranio-maxillo-facial surgery : official publication of the European Association for Cranio-Maxillo-Facial Surgery
The influence of geranylgeraniol on human oral keratinocytes after bisphosphonate treatment: An in vitro study.
PMID 25913629
Abstract
This in vitro study analyzed the influence of geranylgeraniol (GGOH) on human oral keratinocytes (HOK) after exposure to bisphosphonates. HOK were incubated with four different bisphosphonates (clodronate, ibandronate, pamidronate, zoledronate) in two experimental set-ups: with and without GGOH. MTT and PrestoBlue assays were used to analyze HOK cell viability. HOK migration ability was examined with Boyden and Scratch assays, and Tunel and ToxiLight assays were used to detect the HOK apoptosis rate. No significant differences between the experimental set-ups, with and without GGOH, could be found for clodronate (p each >0.3). For the nitrogen-containing bisphosphonates, negative effects could be shown in the experimental set-ups without GGOH in all assays. In the GGOH experimental set-ups, the levels of HOK cell viability were significantly increased (MTT: p each ≤0.001; PrestoBlue: p each ≤0.012). The HOK migration ability was also greater (Boyden: p each <0.001; Scratch: p each ≤0.015). Regarding the apoptosis rate, reduced numbers of apoptotic HOK in the Tunel assay (p each <0.001) and decreased adenylate kinase release in the ToxiLight assay (p each ≤0.002) were observed. GGOH reversed the negative effects of bisphosphonates on HOK. These findings provide evidence that GGOH could be a promising treatment option for BP-ONJ.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G3278
Geranylgeraniol, ≥85% (GC)
C20H34O
424455
Resorufin, Dye content 95 %
C12H7NO3
73144
Resorufin, 95%
C12H7NO3