EMAIL THIS PAGE TO A FRIEND
Nature cell biology
The ITIM-containing receptor LAIR1 is essential for acute myeloid leukaemia development.
PMID 25915125
Abstract
Conventional strategies are not particularly successful in the treatment of leukaemia, and identification of signalling pathways crucial to the activity of leukaemia stem cells will provide targets for the development of new therapies. Here we report that certain receptors containing the immunoreceptor tyrosine-based inhibition motif (ITIM) are crucial for the development of acute myeloid leukaemia (AML). Inhibition of expression of the ITIM-containing receptor LAIR1 does not affect normal haematopoiesis but abolishes leukaemia development. LAIR1 induces activation of SHP-1, which acts as a phosphatase-independent signalling adaptor to recruit CAMK1 for activation of downstream CREB in AML cells. The LAIR1-SHP-1-CAMK1-CREB pathway sustains the survival and self-renewal of AML stem cells. Intervention in the signalling initiated by ITIM-containing receptors such as LAIR1 may result in successful treatment of AML.