EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
The mercury levels in crustaceans and cephalopods from Peninsular Malaysia.
PMID 25916470
Abstract
This study is to determine total mercury in edible tissues of eight species of cephalopods and 12 species of crustaceans purchased from 11 identified major fish landing ports and wet markets throughout Peninsular Malaysia. The concentration of mercury was measured by cold vapor atomic absorption spectrometry (AAS) technique using the Perkin Elmer Flow Injection Mercury System (FIMS-400). In general, the mercury levels were low with concentrations in cephalopods ranging from 0.099 to 2.715xa0mg/kg dry weight (or 0.0184-0.505xa0mg/kg wet weight) and in crustaceans ranging from 0.057 to 1.359xa0mg/kg dry weight (or 0.0111-0.265xa0mg/kg wet weight). The mercury levels showed no significant differences (P > 0.05) between species for both cephalopods and crustaceans. There was no significant correlation between mercury concentrations and the body size of individual for both groups as well. Comparisons with mercury levels obtained found from other previous studies and/or species noted that they were of the same magnitude or relatively low compared to various locations reported worldwide.