EMAIL THIS PAGE TO A FRIEND
Methods (San Diego, Calif.)
Integrated systems for exosome investigation.
PMID 25916618
Abstract
Extracellular vesicles, including exosomes, are currently being investigated to better understand their biogenesis and biological functions. There is also a rapidly growing interest in utilizing exosomes present in patient biofluids for molecular diagnostics in the clinic. Exosomes are natural shuttles of RNA and protein cargo, making them attractive as potential therapeutic delivery vehicles. Here, we describe the methods for using the latest tools and technologies to study exosomes to better understand their roles in cell-to-cell communication, for discovery of clinical biomarkers and to engineer exosomes for therapeutic applications.