EMAIL THIS PAGE TO A FRIEND
Acta tropica
Combined flubendazole-nitazoxanide treatment of cystic echinococcosis: Pharmacokinetic and efficacy assessment in mice.
PMID 25917712
Abstract
The current chemotherapy of cystic echinococcosis (CE) is mainly based on the use of albendazole, and the results have been shown to be highly variable. Thus, new and more efficient treatment options are urgently needed. The goals of the current study were: a) to compare the ex vivo activity of flubendazole (FLBZ) and nitazoxanide (NTZ), given either separately or co-administered, against Echinococcus granulosus protoscoleces and cysts, b) to characterize the plasma disposition kinetics of FLBZ administered alone or combined with NTZ in mice; (c) to compare the in vivo activity of FLBZ and NTZ (either each alone or as a combined treatment) against secondary CE developed in mice. Ex vivo drug activity study: E. granulosus protoscoleces and cysts were incubated either with FLBZ, NTZ, or the FLBZ-NTZ combination. Protoscoleces and cyst viability was monitored by the methylene blue exclusion test and scanning electron microscopy (SEM). Pharmacokinetic study: Balb/C mice received FLBZ (5 mg/kg) orally either alone or co-administered with NTZ (100 mg/kg). Blood samples were collected up to 12 h post treatment and plasma analyzed for FLBZ/metabolites by HPLC. Clinical Efficacy study: following secondary infection, meaning i.p. injection of 1500 E. granulosus protoscoleces/animal (n=40), the both drugs were administered by intragastric inoculation on a daily basis for a period of 25 days. Balb/C mice received FLBZ (5 mg/kg, twice a day) alone, NTZ (100 mg/kg, once daily) alone or a combination of both molecules (FLBZ, 5mg/kg twice a day and NTZ, 100 mg/kg, once daily). Ten untreated animals were used as a control. All animals were killed and the weight of the cysts collected from each animal was recorded. The presence of NTZ did not markedly affect the FLBZ kinetic parameters in mice. FLBZ alone or combined with NTZ induced a reduction (P<0.05) of cyst weight in comparison to the untreated control and NTZ-treated treated mice. The data obtained here indicate that NTZ did not affect hydatid cyst development in mice. Conversely, FLBZ shows an excellent efficacy against CE.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O3132
Oxibendazole, ~98%
C12H15N3O3