EMAIL THIS PAGE TO A FRIEND
Acta tropica
High selective antileishmanial activity of vanadium complex with stilbene derivative.
PMID 25917716
Abstract
Leishmaniasis is a group of disease caused by different species of the parasite Leishmania affecting millions of people worldwide. Conventional therapy relies on multiple parenteral injections with pentavalent antimonials which exhibit high toxicity and various side effects have been reported. Hence, the research for an effective and low toxic effect drug is necessary. In the present work, the synthesis, spectroscopic and analytical characterizations of stilbene derivative (H2Salophen) and its vanadium complex (VOSalophen) are reported. Besides the chemical ancillary information, investigation of the leishmanicidal effects of these compounds were provided. The biological assays against promastigote and amastigote forms of L. amazonensis have been shown that VOSalophen exhibited a strong antiparasitic activity (IC50 of 6.65 and 3.51 μM, respectively). Furthermore, the leishmanicidal activity was concentration and time-dependent. Regarding toxicity and selectivity on mammalian cells, VOSalophen have not caused significant damage to human erythrocytes in all concentrations tested and VOSalophen was almost seven times more destructive for the intracellular parasite than for macrophages. Furthermore, the leishmanicidal activity of VOSalophen in promastigote forms of L. amazonensis could be associated to mitochondrial dysfunction and increase of the reactive oxygen species (ROS) production. In L. amazonensis-infected macrophages, VOSalophen induces ROS production and a microbicidal action NO-dependent. Our biological results indicate the effective and selective action of VOSalophen against L. amazonensis and the leishmanicidal effect can be associated to parasite disorders and immumodulatory effects.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M5571
Miltefosine, ≥98% (perchloric acid titration)
C21H46NO4P
N9125
N-(1-Naphthyl)ethylenediamine dihydrochloride, ≥98%
C12H14N2 · 2HCl
222488
N-(1-Naphthyl)ethylenediamine dihydrochloride, ACS reagent, >98%
C12H14N2 · 2HCl
84160
Salicylaldehyde, redist., ≥99.0% (GC)
C7H6O2
S356
Salicylaldehyde, reagent grade, 98%
C7H6O2
W300403
Salicylaldehyde, ≥98%, FG
C7H6O2
357162
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
V
357170
Vanadium, foil, thickness 0.25 mm, 99.7% trace metals basis
V
266205
Vanadium, foil, thickness 0.5 mm, 99.7% trace metals basis
V
262935
Vanadium, powder, −325 mesh, 99.5% trace metals basis
V
266175
Vanadium, rod, diam. 6.2 mm, 99.7% trace metals basis
V
262927
Vanadium, turnings, 99.7% trace metals basis
V
774073
Vanadium, pieces, 1-3 mm, 99.9% trace metals basis
V
774065
Vanadium, powder, -100 mesh, 99.9% trace metals basis
V
GF98785388
Vanadium, microfoil, disks, 10mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF25800386
Vanadium, wire reel, 5m, diameter 0.5mm, as drawn, 99.8%
V
GF46548692
Vanadium, microfoil, disks, 10mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF21697106
Vanadium, microfoil, disks, 25mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF76957004
Vanadium, microfoil, disks, 25mm, thinness 0.25μm, specific density 140.4μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF23355996
Vanadium, microfoil, disks, 25mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF69763977
Vanadium, wire reel, 0.2m, diameter 0.1mm, as drawn, 99.8%
V
GF75438445
Vanadium, wire reel, 0.2m, diameter 0.25mm, hard, 99.8%
V
GF84027269
Vanadium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.8%
V
GF25047503
Vanadium, wire reel, 0.2m, diameter 1.0mm, Stress relieved, 99.8%
V
GF39723239
Vanadium, wire reel, 0.5m, diameter 0.1mm, as drawn, 99.8%
V
GF29805341
Vanadium, wire reel, 0.5m, diameter 0.25mm, hard, 99.8%
V
GF03408555
Vanadium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.8%
V
GF30937732
Vanadium, wire reel, 0.5m, diameter 1.0mm, Stress relieved, 99.8%
V
GF57656391
Vanadium, wire reel, 1m, diameter 0.1mm, as drawn, 99.8%
V
GF25668344
Vanadium, wire reel, 1m, diameter 0.25mm, hard, 99.8%
V