EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Hygienic regulation of tri(chloroethyl)phosphate in the water].
PMID 2591796
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
119660
Tris(2-chloroethyl) phosphate, 97%
C6H12Cl3O4P