EMAIL THIS PAGE TO A FRIEND
International journal of immunopathology and pharmacology
Effect of carbamate pesticides on perforin, granzymes A-B-3/K, and granulysin in human natural killer cells.
PMID 25921628
Abstract
We previously found that ziram, a carbamate pesticide, significantly reduced perforin, granzyme A (GrA), granzyme B (GrB), granzyme 3/K (Gr3/K), and granulysin (GRN) levels in NK-92MI cells, a human natural killer (NK) cell line. To investigate whether other carbamate pesticides also show similar toxicity on human NK cells, we conducted further experiments with NK-92CI cells, a human NK cell line, using a more sensitive assay. We previously confirmed that NK-92CI cells express CD56, perforin, GrA, GrB, Gr3/K, and GRN and are highly cytotoxic to K562 cells in a chromium release assay, which are more sensitive to organophosphorus pesticides and ziram than the NK-92MI cell line. NK-92CI cells were treated with ziram, thiram, maneb, or carbaryl at various concentrations for 4-24 h at 37°C in vitro. Thereafter, intracellular levels of perforin, GrA, GrB, Gr3/K, and GRN were determined by flow cytometry. It was found that all carbamate pesticides significantly reduced the intracellular levels of perforin, GrA, GrB, Gr3/K, and GRN in NK-92CI cells in a dose-dependent manner. However, the strength of the effect differed among the pesticides, and the order was thiram > ziram > maneb > carbaryl. In addition, it was also found that the degree of the reductions differed among the five proteins, with perforin more sensitive to pesticides than GRN, GrA, GrB, and Gr3/K, and the order was perforin > GRN > Gr3/K ≒ GrA ≒ GrB.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
559814
1-Naphthyl-N-methylcarbamate, 97%
C12H11NO2
329711
Zinc dimethyldithiocarbamate, 97%
C6H12N2S4Zn