EMAIL THIS PAGE TO A FRIEND
Molecular Imaging and Biology : MIB : The Official Publication of the Academy of Molecular Imaging
In Vivo Tracking of Streptococcal Infections of Subcutaneous Origin in a Murine Model.
PMID 25921659